Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)propanoate 95%

BD279674-250MG BD279674-100MG
BLDPBD279674-250MGEA 164 EUR
BLDPBD279674-250MG BLDPBD279674-100MG
Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)propanoate 95%
Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)propanoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₄H₂₃BN₂O₄
MW: 294,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383249
CAS Number: 1220968-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR