Methyl 4-amino-2-methylthiophene-3-carboxylate 97%

BD279806-250MG BD279806-100MG BD279806-1G
BLDPBD279806-250MGEA 176 EUR
BLDPBD279806-250MG BLDPBD279806-100MG BLDPBD279806-1G
Methyl 4-amino-2-methylthiophene-3-carboxylate 97%
Methyl 4-amino-2-methylthiophene-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₉NO₂S
MW: 171,22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01570124
CAS Number: 114943-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR