4-Bromo-6-chloro-2-methylpyridazin-3(2H)-one 95%

BD280331-100MG BD280331-250MG
BLDPBD280331-100MGEA 20 EUR
BLDPBD280331-100MG BLDPBD280331-250MG
4-Bromo-6-chloro-2-methylpyridazin-3(2H)-one 95%
4-Bromo-6-chloro-2-methylpyridazin-3(2H)-one

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₄BrClN₂O
MW: 223,46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21364605
CAS Number: 1178884-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR