2-(4-Methoxyphenyl)isoindoline-1,3-dione 95+%

BD28042-5G BD28042-250MG BD28042-1G BD28042-25G
BLDPBD28042-5GEA 84 EUR
BLDPBD28042-5G BLDPBD28042-250MG BLDPBD28042-1G BLDPBD28042-25G
2-(4-Methoxyphenyl)isoindoline-1,3-dione 95+%
2-(4-Methoxyphenyl)isoindoline-1,3-dione

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₅H₁₁NO₃
MW: 253,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00429499
CAS Number: 2142-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR