trans-tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate 98%

BLDPBD280439-1GEA 199 EUR
BLDPBD280439-1G BLDPBD280439-100MG BLDPBD280439-250MG
trans-tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate 98%
trans-tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₀H₁₉FN₂O₂
MW: 218,27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756122
CAS Number: 907544-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR