Methyl 2-amino-2-methylpropanoate 98%

BD280526-250MG BD280526-100MG BD280526-1G BD280526-25G BD280526-5G
BLDPBD280526-250MGEA 26.3 EUR
BLDPBD280526-250MG BLDPBD280526-100MG BLDPBD280526-1G BLDPBD280526-25G BLDPBD280526-5G
Methyl 2-amino-2-methylpropanoate 98%
Methyl 2-amino-2-methylpropanoate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₁₁NO₂
MW: 117,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12407415
CAS Number: 13257-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR