4-[4-(Hydroxymethyl)phenyl]pyridine 95%

Synonyms: (4-Pyridin-4-ylphenyl)methanol,(4-(Pyridin-4-yl)phenyl)methanol

BD28062-250MG BD28062-100MG BD28062-1G
BLDPBD28062-250MGEA 72.5 EUR
BLDPBD28062-250MG BLDPBD28062-100MG BLDPBD28062-1G
4-[4-(Hydroxymethyl)phenyl]pyridine 95%
4-[4-(Hydroxymethyl)phenyl]pyridine
Formula: C₁₂H₁₁NO
MW: 185,222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114576
CAS Number: 217192-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR