(1R,5S)-tert-Butyl 3,6-diazabicyclo[3.2.0]heptane-6-carboxylate 98%

BLDPBD280731-250MGEA 282 EUR
BLDPBD280731-250MG BLDPBD280731-100MG BLDPBD280731-1G
(1R,5S)-tert-Butyl 3,6-diazabicyclo[3.2.0]heptane-6-carboxylate 98%
(1R,5S)-tert-Butyl 3,6-diazabicyclo[3.2.0]heptane-6-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₀H₁₈N₂O₂
MW: 198,26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22573659
CAS Number: 799279-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR