3-Bromopyridine-N-oxide 95%

Synonyms: 3-Bromopyridine 1-oxide

BLDPBD28134-5GEA 18.9 EUR
BLDPBD28134-5G
3-Bromopyridine-N-oxide 95%
3-Bromopyridine N-oxide

Storage condition: Sealed in dry, 2-8°C

Formula: C₅H₄BrNO
MW: 173,9954 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01646056
CAS Number: 2402-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR