(3-Methoxyphenyl)methanesulphonamide 97%

BD281405-5G BD281405-100MG BD281405-1G BD281405-250MG BD281405-10G
BLDPBD281405-5GEA 491 EUR
BLDPBD281405-5G BLDPBD281405-100MG BLDPBD281405-1G BLDPBD281405-250MG BLDPBD281405-10G
(3-Methoxyphenyl)methanesulphonamide 97%
(3-Methoxyphenyl)methanesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₁NO₃S
MW: 201,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16671946
CAS Number: 89782-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR