Methyl 3-(4-hydroxyphenyl)hex-4-ynoate 95%

BLDPBD282821-250MGEA 64.1 EUR
BLDPBD282821-250MG BLDPBD282821-100MG
Methyl 3-(4-hydroxyphenyl)hex-4-ynoate 95%
Methyl 3-(4-hydroxyphenyl)hex-4-ynoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₃H₁₄O₃
MW: 218,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24713927
CAS Number: 865234-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR