Hydroxy-PEG4-acid 95+%

Synonyms: 1-Hydroxy-3,6,9,12-tetraoxapentadecan-15-oic acid

BD282986-1G BD282986-100MG BD282986-250MG
BLDPBD282986-1GEA 318 EUR
BLDPBD282986-1G BLDPBD282986-100MG BLDPBD282986-250MG
Hydroxy-PEG4-acid 95+%
Hydroxy-PEG4-acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₂₂O₇
MW: 266,288 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22574805
CAS Number: 937188-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR