2-Amino-5-bromo-1,3-thiazole 96%

Supplier: BLD PHARMATECH GMBH

Synonyms: 5-Bromothiazol-2-amine

BLDPBD28318-25GEA 142 EUR
BLDPBD28318-25G BLDPBD28318-1G
2-Amino-5-bromo-1,3-thiazole 96%
2-Amino-5-bromo-1,3-thiazole

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₃H₃BrN₂S
MW: 179,038 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00016893
CAS Number: 3034-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR