1-Chloro-3-(isocyano(tosyl)methyl)benzene 95%

BD28353-250MG BD28353-100MG BD28353-1G
BLDPBD28353-250MGEA 132 EUR
BLDPBD28353-250MG BLDPBD28353-100MG BLDPBD28353-1G
1-Chloro-3-(isocyano(tosyl)methyl)benzene 95%
1-Chloro-3-(isocyano(tosyl)methyl)benzene

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₅H₁₂ClNO₂S
MW: 305,77 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114780
CAS Number: 321345-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR