3-Fluoro-5-methoxypyridin-2-amine 97%

BLDPBD284492-1GEA 801 EUR
BLDPBD284492-1G BLDPBD284492-100MG BLDPBD284492-250MG
3-Fluoro-5-methoxypyridin-2-amine 97%
3-Fluoro-5-methoxypyridin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₇FN₂O
MW: 142,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606158
CAS Number: 1211589-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR