3-Ethylbenzothiazolium bromide 98+%

Synonyms: 3-Ethylbenzo[d]thiazol-3-ium bromide

BLDPBD284722-5GEA 42 EUR
BLDPBD284722-5G BLDPBD284722-100G BLDPBD284722-500G BLDPBD284722-1G BLDPBD284722-25G
3-Ethylbenzothiazolium bromide 98+%
3-Ethylbenzothiazolium bromide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₀BrNS
MW: 244,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051852
CAS Number: 32446-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR