5,6-Dihydroxy-2-phenylpyrimidine-4-carboxylic acid 95%

BLDPBD284889-1GEA 265 EUR
BLDPBD284889-1G BLDPBD284889-250MG BLDPBD284889-100MG
5,6-Dihydroxy-2-phenylpyrimidine-4-carboxylic acid 95%
5,6-Dihydroxy-2-phenylpyrimidine-4-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₈N₂O₄
MW: 232,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09701383
CAS Number: 62222-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR