Methyl 3-(2-oxopropyl)benzoate 95%

BD285117-100MG BD285117-250MG BD285117-1G
BLDPBD285117-100MGEA 311 EUR
BLDPBD285117-100MG BLDPBD285117-250MG BLDPBD285117-1G
Methyl 3-(2-oxopropyl)benzoate 95%
Methyl 3-(2-oxopropyl)benzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₂O₃
MW: 192,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258776
CAS Number: 74998-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR