5-Bromo-3-chloro-1H-indazole 97%

Supplier: BLD PHARMATECH GMBH

BLDPBD28513-1GEA 28.4 EUR
BLDPBD28513-1G BLDPBD28513-10G BLDPBD28513-25G BLDPBD28513-5G
5-Bromo-3-chloro-1H-indazole 97%
5-Bromo-3-chloro-1H-indazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₄BrClN₂
MW: 231,4800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738284
CAS Number: 36760-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR