1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 97%
Supplier: BLD PHARMATECH GMBH
|
|
BLDPBD285174-100MGEA
25.2
EUR
BLDPBD285174-100MG
BLDPBD285174-5G
BLDPBD285174-250MG
BLDPBD285174-1G
BLDPBD285174-25G
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 97%
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₉H₁₄O₄ MW: 186,205 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11847571 CAS Number: 66500-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...