Methyl 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%

BD286019-1G BD286019-100MG BD286019-5G BD286019-250MG
BLDPBD286019-1GEA 185 EUR
BLDPBD286019-1G BLDPBD286019-100MG BLDPBD286019-5G BLDPBD286019-250MG
Methyl 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%
Methyl 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₈BClO₄
MW: 296,55 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846464
CAS Number: 408492-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR