1-(2,2,2-Trifluoroacetyl)indoline-5-sulphonyl chloride 97%

BD286054-250MG BD286054-1G BD286054-100MG
BLDPBD286054-250MGEA 193 EUR
BLDPBD286054-250MG BLDPBD286054-1G BLDPBD286054-100MG
1-(2,2,2-Trifluoroacetyl)indoline-5-sulphonyl chloride 97%
1-(2,2,2-Trifluoroacetyl)indoline-5-sulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₇ClF₃NO₃S
MW: 313,68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09261241
CAS Number: 210691-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR