2-Chloro-7-methoxyquinazoline 95+%

BLDPBD286487-1GEA 223 EUR
BLDPBD286487-1G BLDPBD286487-100MG BLDPBD286487-250MG
2-Chloro-7-methoxyquinazoline 95+%
2-Chloro-7-methoxyquinazoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₇ClN₂O
MW: 194,61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09907933
CAS Number: 953039-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR