3-(4,6-Dichloro-2-(ethoxycarbonyl)-1H-indol-3-yl)acrylic acid 95%

BD286936-250MG BD286936-1G BD286936-100MG BD286936-50MG
BLDPBD286936-250MGEA 369 EUR
BLDPBD286936-250MG BLDPBD286936-1G BLDPBD286936-100MG BLDPBD286936-50MG
3-(4,6-Dichloro-2-(ethoxycarbonyl)-1H-indol-3-yl)acrylic acid 95%
3-(4,6-Dichloro-2-(ethoxycarbonyl)-1H-indol-3-yl)acrylic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₄H₁₁Cl₂NO₄
MW: 328,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114763
CAS Number: 159054-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR