(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-(Hydroxymethyl)-1H-pyrrolo[3,2-b]pyridine

BLDPBD286988-100MGEA 147 EUR
BLDPBD286988-100MG BLDPBD286988-1G BLDPBD286988-250MG
(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol 97%
(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈N₂O
MW: 148,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18432555
CAS Number: 17288-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR