4-Bromo-6-methylpicolinaldehyde 95%

BLDPBD28763-100MGEA 209 EUR
BLDPBD28763-100MG BLDPBD28763-250MG
4-Bromo-6-methylpicolinaldehyde 95%
4-Bromo-6-methylpicolinaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₆BrNO
MW: 200,03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07374948
CAS Number: 448906-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR