(4-Cyanophenyl)acetic acid methyl ester 96%
Supplier: BLD PHARMATECH GMBH
|
|
Synonyms:
Methyl (4-cyanophenyl)acetate
BLDPBD28945-25GEA
480
EUR
BLDPBD28945-25G
BLDPBD28945-10G
BLDPBD28945-100G
BLDPBD28945-5G
BLDPBD28945-250MG
BLDPBD28945-1G
(4-Cyanophenyl)acetic acid methyl ester 96%
(4-Cyanophenyl)acetic acid methyl ester
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₉NO₂ MW: 175,1840 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02181140 CAS Number: 52798-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...