2-(2,6-Difluoro-3-nitrophenyl)acetonitrile 98%

BD289527-1G BD289527-25G BD289527-5G
BLDPBD289527-1GEA 57.8 EUR
BLDPBD289527-1G BLDPBD289527-25G BLDPBD289527-5G
2-(2,6-Difluoro-3-nitrophenyl)acetonitrile 98%
2-(2,6-Difluoro-3-nitrophenyl)acetonitrile

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₄F₂N₂O₂
MW: 198,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22571061
CAS Number: 1369761-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR