(S)-b-Amino-4-methoxy-benzeneethanol 95%
Synonyms:
(S)-2-Amino-2-(4-methoxyphenyl)ethanol
BLDPBD290342-250MGEA
222
EUR
BLDPBD290342-250MG
BLDPBD290342-100MG
(S)-b-Amino-4-methoxy-benzeneethanol 95%
(S)-b-Amino-4-methoxy-benzeneethanol
Formula:
C₉H₁₃NO₂ MW: 167,205 g/mol |
MDL Number:
MFCD09253728 CAS Number: 191109-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...