Methyl 5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate 98%

BLDPBD291127-100MGEA 25.2 EUR
BLDPBD291127-100MG BLDPBD291127-1G BLDPBD291127-5G BLDPBD291127-250MG BLDPBD291127-25G
Methyl 5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate 98%
Methyl 5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₀N₂O₂
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28134587
CAS Number: 1454848-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR