Ethyl-2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: Ethyl 2-(4-methoxyphenyl)thiazole-4-carboxylate

BLDPBD29114-1GEA 43.1 EUR
BLDPBD29114-1G BLDPBD29114-100MG BLDPBD29114-5G BLDPBD29114-250MG
Ethyl-2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate 98%
Ethyl 2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₃NO₃S
MW: 263,3100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738343
CAS Number: 57677-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR