4-Phenyloxazol-2-amine 97%

BD291172-100MG BD291172-1G BD291172-250MG
BLDPBD291172-100MGEA 109 EUR
BLDPBD291172-100MG BLDPBD291172-1G BLDPBD291172-250MG
4-Phenyloxazol-2-amine 97%
4-Phenyloxazol-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₈N₂O
MW: 160,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189375
CAS Number: 33119-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR