Methyl 3-((2,2-dimethylbutanoyl)thio)propanoate 95%

BD291319-1G BD291319-100G BD291319-5G BD291319-10G BD291319-25G
BLDPBD291319-1GEA 6.3 EUR
BLDPBD291319-1G BLDPBD291319-100G BLDPBD291319-5G BLDPBD291319-10G BLDPBD291319-25G
Methyl 3-((2,2-dimethylbutanoyl)thio)propanoate 95%
Methyl 3-((2,2-dimethylbutanoyl)thio)propanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₈O₃S
MW: 218,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22200648
CAS Number: 938063-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR