6-Chloro-1H-indazole 98%

BLDPBD29445-10GEA 133 EUR
BLDPBD29445-10G BLDPBD29445-100MG BLDPBD29445-1G BLDPBD29445-5G BLDPBD29445-250MG
6-Chloro-1H-indazole 98%
6-Chloro-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₅ClN₂
MW: 152,5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774234
CAS Number: 698-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR