1-(5-Chloro-2-pyridinyl)piperazine 98%

Synonyms: 1-(5-Chloro-2-pyridyl)piperazine

BLDPBD29795-1GEA 59.9 EUR
BLDPBD29795-1G BLDPBD29795-25G BLDPBD29795-5G BLDPBD29795-250MG
1-(5-Chloro-2-pyridinyl)piperazine 98%
1-(5-Chloro-2-pyridinyl)piperazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₂ClN₃
MW: 197,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115049
CAS Number: 87394-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR