4-(Aminomethyl)tetrahydropyran 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: (Tetrahydro-2H-pyran-4-yl)methanamine

BLDPBD3006-10GEA 25.2 EUR
BLDPBD3006-10G BLDPBD3006-0.025G BLDPBD3006-25G BLDPBD3006-5G BLDPBD3006-1G
4-(Aminomethyl)tetrahydropyran 97%
4-(Aminomethyl)tetrahydropyran

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₁₃NO
MW: 115,1735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179435
CAS Number: 130290-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR