Methyl 4-bromo-5-chlorothiophene-2-carboxylate 97%

BD303884-250MG BD303884-1G
BLDPBD303884-250MGEA 10.5 EUR
BLDPBD303884-250MG BLDPBD303884-1G
Methyl 4-bromo-5-chlorothiophene-2-carboxylate 97%
Methyl 4-bromo-5-chlorothiophene-2-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₄BrClO₂S
MW: 255,52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28359275
CAS Number: 1047630-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR