1-(4-Bromophenyl)-3-methylthiourea 97%

BD303886-1G BD303886-250MG BD303886-100MG
BLDPBD303886-1GEA 198 EUR
BLDPBD303886-1G BLDPBD303886-250MG BLDPBD303886-100MG
1-(4-Bromophenyl)-3-methylthiourea 97%
1-(4-Bromophenyl)-3-methylthiourea

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₉BrN₂S
MW: 245,13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02940754
CAS Number: 61449-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR