(3-Methyl-4-morpholinophenyl)boronic acid 97%

BD303891-250MG BD303891-5G BD303891-1G
BLDPBD303891-250MGEA 45.2 EUR
BLDPBD303891-250MG BLDPBD303891-5G BLDPBD303891-1G
(3-Methyl-4-morpholinophenyl)boronic acid 97%
(3-Methyl-4-morpholinophenyl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₁₆BNO₃
MW: 221,06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16495874
CAS Number: 1426245-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR