(5-Methylthiophen-3-yl)boronic acid 95+%

BD303955-250MG BD303955-100MG
BLDPBD303955-250MGEA 69.3 EUR
BLDPBD303955-250MG BLDPBD303955-100MG
(5-Methylthiophen-3-yl)boronic acid 95+%
(5-Methylthiophen-3-yl)boronic acid

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formula: C₅H₇BO₂S
MW: 141,98 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18250237
CAS Number: 930303-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR