4-Methyl-2-(piperidin-1-yl)quinoline 98%

Synonyms: ML 204

BD303983-250MG BD303983-100MG
BLDPBD303983-250MGEA 198 EUR
BLDPBD303983-250MG BLDPBD303983-100MG
4-Methyl-2-(piperidin-1-yl)quinoline 98%
4-Methyl-2-(piperidin-1-yl)quinoline

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₅H₁₈N₂
MW: 226,32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00817920
CAS Number: 5465-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR