3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%

BD30404-5G BD30404-1G BD30404-250MG BD30404-10G BD30404-25G
BLDPBD30404-5GEA 58.8 EUR
BLDPBD30404-5G BLDPBD30404-1G BLDPBD30404-250MG BLDPBD30404-10G BLDPBD30404-25G
3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%
3-Amino-2-chloro-6-(trifluoromethyl)pyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C6H4ClF3N2
MW: 196,5600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862661
CAS Number: 117519-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR