N-(2-(2-Methyl-1H-indol-3-yl)ethyl)thiophene-2-carboxamide 98%

BD304052-100MG BD304052-250MG BD304052-1G
BLDPBD304052-100MGEA 103 EUR
BLDPBD304052-100MG BLDPBD304052-250MG BLDPBD304052-1G
N-(2-(2-Methyl-1H-indol-3-yl)ethyl)thiophene-2-carboxamide 98%
N-(2-(2-Methyl-1H-indol-3-yl)ethyl)thiophene-2-carboxamide

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₆H₁₆N₂OS
MW: 284,37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03036245
CAS Number: 442632-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR