(R)-4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one 98%

BD304071-100MG BD304071-1G BD304071-5G BD304071-250MG
BLDPBD304071-100MGEA 47.3 EUR
BLDPBD304071-100MG BLDPBD304071-1G BLDPBD304071-5G BLDPBD304071-250MG
(R)-4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one 98%
(R)-4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₆H₁₃FO
MW: 240,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28963660
CAS Number: 1234356-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR