4-(2,2,2-Trifluoroethoxy)benzoic acid 97%

BD304076-5G BD304076-1G BD304076-100MG BD304076-250MG
BLDPBD304076-5GEA 135 EUR
BLDPBD304076-5G BLDPBD304076-1G BLDPBD304076-100MG BLDPBD304076-250MG
4-(2,2,2-Trifluoroethoxy)benzoic acid 97%
4-(2,2,2-Trifluoroethoxy)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₇F₃O₃
MW: 220,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05669500
CAS Number: 27914-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR