ent-Cinacalcet Hydrochloride 97%

BD304099-5G BD304099-1G BD304099-250MG BD304099-100MG
BLDPBD304099-5GEA 1010 EUR
BLDPBD304099-5G BLDPBD304099-1G BLDPBD304099-250MG BLDPBD304099-100MG
ent-Cinacalcet Hydrochloride 97%
ent-Cinacalcet Hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₂H₂₃ClF₃N
MW: 393,87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23160290
CAS Number: 1217809-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR