Ethyl 6-Fluoro-1,2,3,4-tetrahydro-isoquinoline-1-carboxylate hydrochloride 95%

Synonyms: Ethyl 6-fluoro-1,2,3,4-tetrahydroisoquinoline-1-carboxylate hydrochloride

BD304114-100MG
BLDPBD304114-100MGEA 38.9 EUR
BLDPBD304114-100MG
Ethyl 6-Fluoro-1,2,3,4-tetrahydro-isoquinoline-1-carboxylate hydrochloride 95%
Ethyl 6-Fluoro-1,2,3,4-tetrahydro-isoquinoline-1-carboxylate hydrochloride
Formula: C₁₂H₁₄FNO₂·ClH
MW: 259,704 g/mol
MDL Number: MFCD28155351
CAS Number: 1822454-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR