8-Methoxyquinoline-4-carbaldehyde 95%

BD304186-250MG BD304186-1G BD304186-100MG
BLDPBD304186-250MGEA 126 EUR
BLDPBD304186-250MG BLDPBD304186-1G BLDPBD304186-100MG
8-Methoxyquinoline-4-carbaldehyde 95%
8-Methoxyquinoline-4-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₉NO₂
MW: 187,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06824369
CAS Number: 103854-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR