1-Bromo-2,6-bis(trifluoromethyl)benzene 98%

Synonyms: 2-Bromo-1,3-bis(trifluoromethyl)benzene,1,3-Bis(trifluoromethyl)-2-bromobenzene

BD30419-250MG BD30419-5G BD30419-25G BD30419-10G BD30419-1G BD30419-100G
BLDPBD30419-250MGEA 6.3 EUR
BLDPBD30419-250MG BLDPBD30419-5G BLDPBD30419-25G BLDPBD30419-10G BLDPBD30419-1G BLDPBD30419-100G
1-Bromo-2,6-bis(trifluoromethyl)benzene 98%
1-Bromo-2,6-bis(trifluoromethyl)benzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₃BrF₆
MW: 293,0038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236210
CAS Number: 118527-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR