3-Chloroquinoline-7-carboxylic acid 95%

BD304202-100MG BD304202-250MG
BLDPBD304202-100MGEA 381 EUR
BLDPBD304202-100MG BLDPBD304202-250MG
3-Chloroquinoline-7-carboxylic acid 95%
3-Chloroquinoline-7-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₆ClNO₂
MW: 207,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28133479
CAS Number: 1824051-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR